Mercury(II) acetate ≥98.0% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Mercury diacetate, Mercuric acetate, Acetic acid mercury (II) salt

12273-18 12273-30
CAAA12273-18EA 81.84 CAD
CAAA12273-18 CAAA12273-30
Mercury(II) acetate ≥98.0% ACS
Mercury (II) acetate

MDL: MFCD00012165 Beilstein Registry No.: 3563831 Fieser: 1,644 6,358 10,252 15,198 17,176 18,232 19,211 20,238 21,263


Purity: 98.0% min CAS No.: 1600-27-7 Molecular Formula: C4H6HgO4 Formula Weight: 318.68 Physical Form: Crystalline Density: 3.28 Appearance: White Odor: Weak Melting Point: 179-182° Beilstein Reference: 3563831 Merck Reference: 14,5873 UN No.: 1629 MDL No.: MFCD00012165

Formula: (H₃CCOO)₂Hg
MW: 318.68 g/mol
Melting Pt: 179…182 °C
Density: 3.28
Storage Temperature: Ambient
MDL Number: MFCD00012165
CAS Number: 1600-27-7
EINECS: 216-491-1
UN: 1629
ADR: 6.1 II
Merck Index: 14,5873

Order Now

Specification Test Results

Insoluble matter 0.01%
Residue after reduction 0.02%
Cl 0.005%
NO3 0.005%
SO3 0.005%
Heavy Metals (as Pb) 0.002%
Fe 0.001%
Mercurous mercury (as Hg) 0.4%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR