Gallium phosphide ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
12266-04
12266-09
12266-18
CAAA12266-04EA
240.16
CAD
CAAA12266-04
CAAA12266-09
CAAA12266-18
Gallium phosphide ≥99.999% (metals basis)
Gallium phosphide
In semiconducting devices
Purity: 99.999%
CAS No.: 12063-98-8
Molecular Formula: GaP
Formula Weight: 100.69
Physical Form: 6mm & Down Pieces
Density: 4.13
Appearance: Pale orange
Melting Point: 1465°
Merck Reference: 14,4353
MDL No.: MFCD00016109
In semiconducting devices
Formula:
GaP Melting Pt: 1465 °C Density: 4.13 Storage Temperature: Ambient |
MDL Number:
MFCD00016109 CAS Number: 12063-98-8 EINECS: 235-057-2 Merck Index: 14,4353 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...