Terbium ≥99.9% (REO, rare earth oxide basis), pieces, distilled dendritic

Supplier: Thermo Scientific Chemicals
Danger

12258-03 12258-06 12258-14
CAAA12258-03EA 131.14 CAD
CAAA12258-03 CAAA12258-06 CAAA12258-14
Terbium ≥99.9% (REO, rare earth oxide basis), pieces, distilled dendritic
Terbium

MDL: MFCD00011256


Purity: 99.9% CAS No.: 7440-27-9 Physical Form: Packaged under argon Appearance: Silver-grey Odor: Odorless UN No.: 3178 MDL No.: MFCD00011256

Formula: Tb
Boiling Pt: 3041 °C (1013 hPa)
Melting Pt: 1356 °C
Density: 8.27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011256
CAS Number: 7440-27-9
EINECS: 231-137-6
UN: 3178
ADR: 4.1,II
Merck Index: 12,09300

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR