Dysprosium ≥99.9% (REO, rare earth oxide basis), pieces, distilled dendritic

Supplier: Thermo Scientific Chemicals
Danger

12257-06 12257-14 12257-22
CAAA12257-06EA 141.25 CAD
CAAA12257-06 CAAA12257-14 CAAA12257-22
Dysprosium ≥99.9% (REO, rare earth oxide basis), pieces, distilled dendritic
Dysprosium

MDL: MFCD00010982


Purity: 99.9% CAS No.: 7429-91-6 Physical Form: Packaged under argon Appearance: Silver Odor: Odorless UN No.: 3178 MDL No.: MFCD00010982

Formula: Dy
Storage Temperature: Ambient
MDL Number: MFCD00010982
CAS Number: 7429-91-6
EINECS: 231-073-9
UN: 3178
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR