Vanadium ≥99.5% (metals basis), powder -325 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
12234-03
12234-09
12234-18
12234-30
CAAA12234-03EA
48.35
CAD
CAAA12234-03
CAAA12234-09
CAAA12234-18
CAAA12234-30
Vanadium ≥99.5% (metals basis), powder -325 mesh
Vanadium
MDL: MFCD00011453
Purity: 99.5%
CAS No.: 7440-62-2
Physical Form: Powder
Density: 6.11
Appearance: Pale gray
Odor: Odorless
Boiling Point: 3000°
UN No.: 3089
MDL No.: MFCD00011453
Formula:
V Boiling Pt: 3000 °C Melting Pt: 1900 °C Density: 6,11 Storage Temperature: Ambient |
MDL Number:
MFCD00011453 CAS Number: 7440-62-2 EINECS: 231-171-1 UN: 3089 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...