Bismuth(III) nitrate pentahydrate ≥98% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Application in luminous paints, preparation of bismuth salts
Purity: 98%
CAS No.: 10035-06-0
Molecular Formula: Bi(NO3)3.5H2O
Formula Weight: 485.07 (394.99anhy)
Physical Form: Crystalline
Density: 2.83
Melting Point: 30° dec.
Solubility: Soluble in dilute HNO3 acid solution. Decomposes in water to subnitrate. Soluble in dilute acetic acid, glycerol, acetone. Insoluble in alcohol, ethyl acetate
Merck Reference: 14,1271
UN No.: 1477
MDL No.: MFCD00149157
Luminous paints, preparation of bismuth salts
Formula:
Bi(NO₃)₃·5H₂O Boiling Pt: 75…80 °C (1013 hPa) Melting Pt: 30 °C (dec.) Density: 2.83 Storage Temperature: Ambient |
MDL Number:
MFCD00149157 CAS Number: 10035-06-0 EINECS: 233-791-8 UN: 1477 ADR: 5.1,II Merck Index: 14,1271 |
Specification Test Results
Insoluble matter | 0.005% |
Cl | 0.1% |
SO3 | 0.005% |
As | 0.00001 |
Cu | 0.002% |
Fe | 0.001% |
PB | 0.002% |
K | 0.01% |
NA | 0.02% |
Ag | 0.001% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...