Cadmium(II) nitrate tetrahydrate ≥98.5%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Manufacturing of Cd salts, photographic emulsions, laboratory reagent, coloring glass
Purity: 98.5% min
CAS No.: 10022-68-1
Molecular Formula: Cd(NO3)2.4H2O
Formula Weight: 308.47 (236.41anhy)
Physical Form: Crystalline
Density: 2.45
Melting Point: 59°
Boiling Point: 132°
Solubility: Soluble in water, alcohol, acetone, ethyl acetate. Insoluble in HNO3
Merck Reference: 14,1622
UN No.: 3087
MDL No.: MFCD00149626
Manufacturing of Cd salts, photographic emulsions, laboratory reagent, coloring glass
Formula:
CdN₂O₆·4H₂O Boiling Pt: 132 °C Melting Pt: 59 °C Density: 2.45 Storage Temperature: Ambient |
MDL Number:
MFCD00149626 CAS Number: 10022-68-1 EINECS: 233-710-6 UN: 3087 ADR: 5.1,II Merck Index: 14,1622 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...