Cobalt(II) acetate tetrahydrate Co 24%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cobaltous acetate tetrahydrate, Acetic acid cobalt (II) salt tetrahydrate, Cobalt(Ⅱ) acetate tetrahydrate, Cobalt diacetate tetrahydrate
MDL: MFCD00149569
Beilstein Registry No.: 3731397
Soluble in water, alcohol, dilute acids
Fieser: 18,106 20,115
CAS No.: 6147-53-1
Molecular Formula: C4H6CoO4.4H2O
Formula Weight: 249.08 (177.02anhy)
Physical Form: Crystals
Density: 1.71
Refractive Index: 1.542
Sensitivity: Hygroscopic
Appearance: Red
Odor: Odorless
Melting Point: ca 140° -4H2O
Solubility: Soluble in water, alcohol, dilute acids
Beilstein Reference: 3731397
Merck Reference: 14,2433
UN No.: 3077
MDL No.: MFCD00149569
Formula:
(H₃CCOO)₂Co·4H₂O MW: 249.08 g/mol Melting Pt: ∼140 °C (-4H₂O) Density: 1.71 Storage Temperature: Ambient |
MDL Number:
MFCD00149569 CAS Number: 6147-53-1 EINECS: 200-755-8 UN: 3077 ADR: 9,III Merck Index: 14,2433 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...