Iron(III) nitrate nonahydrate 98+% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ferric nitrate nonahydrate, Iron trinitrate nonahydrate, Iron(III) nitrate hydrate, Ferric Nitrate, 9-Hydrate

12226-22 12226-A1 12226-A7
CAAA12226-22EA 54.35 CAD
CAAA12226-22 CAAA12226-A1 CAAA12226-A7
Iron(III) nitrate nonahydrate 98+% (metals basis)
Iron (III) nitrate nonahydrate

MDL: MFCD00149708


Purity: 98+% CAS No.: 7782-61-8 Molecular Formula: FeN3O9.9H2O Formula Weight: 403.99 (241.86anhy) Physical Form: Crystalline, Contains excess HNO3 Density: 1.684 Appearance: Pale purple Odor: Acidic Melting Point: 47° Boiling Point: 125° dec. Merck Reference: 14,4027 UN No.: 1466 MDL No.: MFCD00149708

Formula: Fe(NO₃)₃·9H₂O
MW: 404 g/mol
Boiling Pt: 125 °C (dec.)
Melting Pt: 47 °C
Density: 1.684
Storage Temperature: Ambient
MDL Number: MFCD00149708
CAS Number: 7782-61-8
EINECS: 233-899-5
UN: 1466
ADR: 5.1,III
Merck Index: 14,4027

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR