Magnesium acetate tetrahydrate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Acetic acid magnesium salt tetrahydrate, Magnesium di(acetate) tetrahydrate

12225-30 12225-A1
CAAA12225-30EA 89.43 CAD
CAAA12225-30 CAAA12225-A1
Magnesium acetate tetrahydrate 98%
Magnesium acetate tetrahydrate

MDL: MFCD00149214 Beilstein Registry No.: 3730605 Freely soluble in water, alcohol


Purity: 98% CAS No.: 16674-78-5 Molecular Formula: Mg(CH3COO)2.4H2O Formula Weight: 214.45 (142.39anhy) Physical Form: Crystalline Density: 1.454 Melting Point: 80° Solubility: Freely soluble in water, alcohol Beilstein Reference: 3730605 Merck Reference: 14,5654 MDL No.: MFCD00149214

Formula: (H₃CCOO)₂Mg·4H₂O
MW: 214.46 g/mol
Melting Pt: 80 °C
Density: 1.454
Storage Temperature: Ambient
MDL Number: MFCD00149214
CAS Number: 16674-78-5
EINECS: 205-554-9
Merck Index: 14,5654

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR