Nickel acetate hydrate 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nickel(II) acetate hydrate

12223-30 12223-A1 12223-A7
CAAA12223-30EA 80.26 CAD
CAAA12223-30 CAAA12223-A1 CAAA12223-A7
Nickel acetate hydrate 99+%
Nickel acetate hydrate

MDL: MFCD00012451 Soluble in water, alcohol Fieser: 20,254


Purity: 99+% CAS No.: 6018-89-9 Molecular Formula: Ni(CH3COO)2.xH2O Formula Weight: 176.79(anhy) Physical Form: Crystalline Appearance: Green Odor: Light Melting Point: dec. Solubility: Soluble in water, alcohol Merck Reference: 14,6499 UN No.: 3077 MDL No.: MFCD00012451

Formula: C₄H₆NiO₄·H₂O
Melting Pt: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00012451
CAS Number: 6018-89-9
EINECS: 206-761-7
UN: 3077
ADR: 9,III
Merck Index: 14,6499

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR