Nickel(II) nitrate hexahydrate Ni ≥ 19.8%, tech.
Supplier: Thermo Scientific Chemicals
|
Danger
|
12222-36
12222-A3
CAAA12222-36EA
85.48
CAD
CAAA12222-36
CAAA12222-A3
Nickel(II) nitrate hexahydrate Ni ≥ 19.8%, tech.
Nickel (II) nitrate hexahydrate
Nickel plating, manufacturing of ceramic colors
CAS No.: 13478-00-7
Molecular Formula: N2NiO6.6H2O
Formula Weight: 290.80 (182.70anhy)
Physical Form: Flake
Density: 2.05
Sensitivity: Hygroscopic
Appearance: Green
Odor: Odorless
Melting Point: 56.7°
Boiling Point: 137°
Solubility: Soluble in water, alcohol
Merck Reference: 14,6513
UN No.: 2725
MDL No.: MFCD00149805
Nickel plating, manufacturing of ceramic colors
Formula:
Ni(NO₃)₂·6H₂O Boiling Pt: 137 °C Melting Pt: 56.7 °C Density: 2.05 Storage Temperature: Ambient |
MDL Number:
MFCD00149805 CAS Number: 13478-00-7 EINECS: 236-068-5 UN: 2725 ADR: 5.1,III Merck Index: 14,6513 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...