Lead(II) oxide ≥99.9%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lead monoxide

12220-36 12220-A3
CAAA12220-36EA 97.8 CAD
CAAA12220-36 CAAA12220-A3
Lead(II) oxide ≥99.9%
Lead (II) oxide

MDL: MFCD00011164


Purity: 99.9% CAS No.: 1317-36-8 Molecular Formula: PbO Formula Weight: 223.19 Physical Form: -325 Mesh Powder Density: 9.53 Melting Point: 886° Merck Reference: 14,5413 UN No.: 3077 MDL No.: MFCD00011164

Formula: PbO
MW: 223.2 g/mol
Boiling Pt: 1470
Melting Pt: 886 °C
Density: 9.53
Storage Temperature: Ambient
MDL Number: MFCD00011164
CAS Number: 1317-36-8
EINECS: 215-267-0
UN: 3077
ADR: 9 III
Merck Index: 14,5413

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR