Cadmium oxide ≥99.95% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

12219-36 12219-A3
CAAA12219-36EA 195.92 CAD
CAAA12219-36 CAAA12219-A3
Cadmium oxide ≥99.95% (metals basis)
Cadmium oxide

MDL: MFCD00010921


Purity: 99.95% CAS No.: 1306-19-0 Molecular Formula: CdO Formula Weight: 128.40 Physical Form: -325 Mesh Powder Density: 8.15 Appearance: Red-brown Odor: Odorless Melting Point: 1430° Merck Reference: 14,1623 UN No.: 2570 MDL No.: MFCD00010921

Formula: CdO
Boiling Pt: 1559 °C (1013 hPa)
Melting Pt: 1430 °C
Density: 8.15
Storage Temperature: Ambient
MDL Number: MFCD00010921
CAS Number: 1306-19-0
EINECS: 215-146-2
UN: 2570
ADR: 6.1,III
Merck Index: 14,1623

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR