Molybdenum(IV) sulfide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Molybdenum disulfide, Molybdenum disulphide

12213-30 12213-A1
CAAA12213-30EA 170.64 CAD
CAAA12213-30 CAAA12213-A1
Molybdenum(IV) sulfide 98%
Molybdenum (IV) sulphide

Hydrogenation and isomerization catalyst, dry lubricant


Purity: 98% CAS No.: 1317-33-5 Molecular Formula: MoS2 Formula Weight: 160.08 Physical Form: Powder Appearance: Black Odor: Odorless Melting Point: 1185° Solubility: Insoluble in water and dilute acids Merck Reference: 14,6236 MDL No.: MFCD00003470


Hydrogenation and isomerization catalyst, dry lubricant

Formula: MoS₂
MW: 160.07 g/mol
Melting Pt: 1185 °C
Density: 4.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003470
CAS Number: 1317-33-5
EINECS: 215-263-9
Merck Index: 14,6236

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR