Bismuth ≥99.99% (metals basis), pieces ≤ 10 cm (3.9 in)

Supplier: Thermo Scientific Chemicals

12208-30 12208-A1 12208-A7
CAAA12208-30EA 148.84 CAD
CAAA12208-30 CAAA12208-A1 CAAA12208-A7
Bismuth ≥99.99% (metals basis), pieces ≤ 10 cm (3.9 in)
Bismuth

MDL: MFCD00134033


Purity: 99.99% CAS No.: 7440-69-9 Odor: Odorless MDL No.: MFCD00134033

Formula: Bi
Boiling Pt: 1490 °C (1013 hPa)
Melting Pt: 271…272 °C
Density: 9.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
Merck Index: 12,01297

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR