Strontium chloride, anhydrous ≥95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
12202-36
12202-A3
CAAA12202-36EA
114.87
CAD
CAAA12202-36
CAAA12202-A3
Strontium chloride, anhydrous ≥95%
Strontium chloride
MDL: MFCD00011249
Purity: 95%
CAS No.: 10476-85-4
Molecular Formula: SrCl2
Formula Weight: 158.53
Physical Form: Powder
Density: 3.052
Refractive Index: 1.650
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 873°
Boiling Point: 1250°
Merck Reference: 14,8840
MDL No.: MFCD00011249
Formula:
Cl₂Sr Boiling Pt: 1250 °C Melting Pt: 873 °C Density: 3.052 Storage Temperature: Ambient |
MDL Number:
MFCD00011249 CAS Number: 10476-85-4 EINECS: 233-971-6 Merck Index: 14,8840 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...