Strontium fluoride ≥99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
12201-30
12201-A1
12201-A7
CAAA12201-30EA
90.38
CAD
CAAA12201-30
CAAA12201-A1
CAAA12201-A7
Strontium fluoride ≥99%
Strontium fluoride
MDL: MFCD00011251
Slightly soluble in water (11.7mg at 18°C/100ml), dilute acids. Decomposes in strong acids.
Purity: 99% min
CAS No.: 7783-48-4
Molecular Formula: SrF2
Formula Weight: 125.62
Physical Form: Powder
Density: 4.240
Refractive Index: 1.442
Appearance: White
Odor: Odorless
Melting Point: 1473°
Solubility: Slightly soluble in water (11.7mg at 18°C/100ml), dilute acids. Decomposes in strong acids
Merck Reference: 14,8842
MDL No.: MFCD00011251
Formula:
F₂Sr Boiling Pt: 2460 °C (1013 hPa) Melting Pt: 1473 °C Density: 4.240 Storage Temperature: Ambient |
MDL Number:
MFCD00011251 CAS Number: 7783-48-4 EINECS: 232-000-3 UN: 3288 ADR: 6.1,III Merck Index: 14,8842 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...