Strontium hydroxide octahydrate, tech.
Supplier: Thermo Scientific Chemicals
|
Danger
|
12199-18
12199-A3
12199-A9
CAAA12199-18EA
45.66
CAD
CAAA12199-18
CAAA12199-A3
CAAA12199-A9
Strontium hydroxide octahydrate, tech.
Strontium hydroxide octahydrate
MDL: MFCD00150395
CAS No.: 1311-10-0
Molecular Formula: Sr(OH)2.8H2O
Formula Weight: 265.76 (121.60anhy)
Physical Form: Crystalline
Density: 1.900
Sensitivity: Air Sensitive and Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 100° -8H2O
Merck Reference: 14,8843
UN No.: 3262
MDL No.: MFCD00150395
Formula:
H₂O₂Sr·8H₂O Melting Pt: 100 °C (-8H₂O) Density: 1.900 Storage Temperature: Ambient |
MDL Number:
MFCD00150395 CAS Number: 1311-10-0 EINECS: 242-367-1 UN: 3262 ADR: 8,III Merck Index: 14,8843 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...