Barium acetate 99.0-102.0% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Barium di(acetate), Acetic acid barium salt

12198-30 12198-A1 12198-A7
CAAA12198-30EA 109.18 CAD
CAAA12198-30 CAAA12198-A1 CAAA12198-A7
Barium acetate 99.0-102.0% ACS
Barium acetate

MDL: MFCD00012447


Purity: 99.0-102.0% CAS No.: 543-80-6 Molecular Formula: Ba(OOCCH3)2 Formula Weight: 255.43 Physical Form: Crystalline Density: 2.47 Merck Reference: 14,965 UN No.: 1564 MDL No.: MFCD00012447

Formula: (H₃CCOO)₂Ba
MW: 255.42 g/mol
Melting Pt: 450
Density: 2.47
Storage Temperature: Ambient
MDL Number: MFCD00012447
CAS Number: 543-80-6
EINECS: 208-849-0
UN: 1564
ADR: 6.1 III
Merck Index: 14,965

Order Now

Specification Test Results

Insoluble matter 0.01%
Cl 0.1%
Oxidizing substances (as NO3) 0.005%
K 0.003%
NA 0.005%
Ca 0.05%
Heavy Metals (as Pb) 5 ppm max.
Fe 0.001%
Sr Pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR