Barium bromide dihydrate ≥99.3%

Supplier: Thermo Scientific Chemicals
Warning

12197-22 12197-36
CAAA12197-22EA 99.38 CAD
CAAA12197-22 CAAA12197-36
Barium bromide dihydrate ≥99.3%
Barium bromide dihydrate

MDL: MFCD00149153 Notes: Loses 1H2O at 75°C and 2H2O at 120°C Very soluble in water, soluble in methanol


Purity: 99.3% CAS No.: 7791-28-8 Molecular Formula: BaBr2.2H2O Formula Weight: 333.17 (297.14anhy) Physical Form: Crystalline Density: 3.58 Appearance: White Solubility: Very soluble in water, soluble in methanol Merck Reference: 14,968 UN No.: 1564 MDL No.: MFCD00149153

Formula: BaBr₂·2H₂O
MW: 333.17 g/mol
Melting Pt: 850 °C
Density: 3.58
Storage Temperature: Ambient
MDL Number: MFCD00149153
CAS Number: 7791-28-8
EINECS: 234-140-0
UN: 1564
ADR: 6.1,III
Merck Index: 14,968

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR