Barium hydroxide, anhydrous 94-98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
12195-18
12195-30
12195-A1
CAAA12195-18EA
45.35
CAD
CAAA12195-18
CAAA12195-30
CAAA12195-A1
Barium hydroxide, anhydrous 94-98%
Barium hydroxide
MDL: MFCD00003443
Purity: 94-98%
CAS No.: 17194-00-2
Molecular Formula: Ba(OH)2
Formula Weight: 171.36
Physical Form: Powder
Sensitivity: Air Sensitive and Hygroscopic
Appearance: White
Odor: Odorless
pH: 12.5
Melting Point: 408°
Merck Reference: 14,977
UN No.: 3262
MDL No.: MFCD00003443
Formula:
H₂BaO₂ Melting Pt: 408 °C Density: 2.18 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003443 CAS Number: 17194-00-2 EINECS: 241-234-5 UN: 3262 ADR: 8,III Merck Index: 14,977 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...