Dipotassium hexachloroosmate Os ≥ 38.7%
Supplier: Thermo Scientific Chemicals
|
Warning
|
12177-01
12177-03
12177-06
CAAA12177-01EA
131.14
CAD
CAAA12177-01
CAAA12177-03
CAAA12177-06
Dipotassium hexachloroosmate Os ≥ 38.7%
Dipotassium hexachloroosmate
MDL: MFCD00011371
Soluble in water. Sparingly soluble in alcohol
CAS No.: 16871-60-6
Molecular Formula: K2Cl6Os
Formula Weight: 481.12
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: Dark red
Melting Point: dec.
Solubility: Soluble in water. Sparingly soluble in alcohol
Merck Reference: 14,7635
MDL No.: MFCD00011371
Formula:
Cl₆K₂Os Melting Pt: Dec. Density: 3.42 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011371 CAS Number: 16871-60-6 EINECS: 240-893-6 Merck Index: 14,7635 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...