Platinum(II) bromide (≥54,5% Pt) ≥99.9% (metals basis) Pt ≥ 54.5%
Supplier: Thermo Scientific Chemicals
|
Warning
|
12172-03
12172-06
CAAA12172-03EA
222.78
CAD
CAAA12172-03
CAAA12172-06
Platinum(II) bromide (≥54,5% Pt) ≥99.9% (metals basis) Pt ≥ 54.5%
Platinum (II) bromide
MDL: MFCD00011180
Purity: 99.9%
CAS No.: 13455-12-4
Molecular Formula: PtBr2
Formula Weight: 354.91
Physical Form: Crystalline
Melting Point: 250° dec.
MDL No.: MFCD00011180
Formula:
Br₂Pt Melting Pt: 250 °C (dec.) Density: 6,652 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011180 CAS Number: 13455-12-4 EINECS: 236-648-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...