Platinum(II) iodide ≥99.99% (metals basis) Pt ≥ 43.0%, Premion®

Supplier: Thermo Scientific Chemicals
Warning

12170-01 12170-03 12170-06
CAAA12170-01EA 79.32 CAD
CAAA12170-01 CAAA12170-03 CAAA12170-06
Platinum(II) iodide ≥99.99% (metals basis) Pt ≥ 43.0%, Premion®
Platinum (II) iodide

MDL: MFCD00011183 Insoluble in water, alkali iodides


Purity: 99.99% CAS No.: 7790-39-8 Molecular Formula: PtI2 Formula Weight: 448.90 Physical Form: Powder Odor: Odorless MDL No.: MFCD00011183

Formula: I₂Pt
Melting Pt: 325 °C (dec.)
Density: 6.403 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011183
CAS Number: 7790-39-8
EINECS: 232-204-2
Merck Index: 12,07688

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR