Potassium hexachloroplatinate(IV) Pt 39.6%
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00011389
Soluble in hot water. Slightly soluble in cold water. Practically insoluble in alcohol
CAS No.: 16921-30-5
Molecular Formula: Cl6K2Pt
Formula Weight: 486.01
Physical Form: Crystalline powder
Density: 3.50
Refractive Index: ≈1.825
Sensitivity: Hygroscopic
Appearance: Yellow
Odor: Odorless
Melting Point: 250°dec.
Solubility: Soluble in hot water. Slightly soluble in cold water. Practically insoluble in alcohol
Merck Reference: 14,7636
UN No.: 3288
MDL No.: MFCD00011389
Formula:
K₂PtCl₆ Melting Pt: 250 °C. (dec.) Density: 3.50 Storage Temperature: Ambient |
MDL Number:
MFCD00011389 CAS Number: 16921-30-5 EINECS: 240-979-3 UN: 3288 ADR: 6.1,III Merck Index: 14,7636 |
Specification Test Results
Form | Crystalline powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...