Sodium tetrachloroaurate dihydrate ≥99.99% (metals basis) Au 49-50%, Premion®
Supplier: Thermo Scientific Chemicals
|
Danger
|
12148-03
12148-06
CAAA12148-03EA
290.72
CAD
CAAA12148-03
CAAA12148-06
Sodium tetrachloroaurate dihydrate ≥99.99% (metals basis) Au 49-50%, Premion®
Sodium tetrachloroaurate dihydrate
MDL: MFCD00149162
Soluble in water, alcohol, ether
Purity: 99.99%
CAS No.: 13874-02-7
Molecular Formula: AuCl4Na.2H2O
Formula Weight: 397.80 (361.77anhy)
Physical Form: Crystalline
Density: 0.8
Appearance: Yellow-orange
Melting Point: 100° dec.
Solubility: Soluble in water, alcohol, ether
Merck Reference: 14,8688
UN No.: 3260
MDL No.: MFCD00149162
Formula:
NaAuCl₄·2H₂O Melting Pt: 100 °C (dec.) Density: 0.8 Storage Temperature: Ambient |
MDL Number:
MFCD00149162 CAS Number: 13874-02-7 EINECS: 239-241-3 UN: 3260 ADR: 8,III Merck Index: 14,8688 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...