Tantalum carbide ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
12144-09
12144-18
12144-30
CAAA12144-09EA
131.46
CAD
CAAA12144-09
CAAA12144-18
CAAA12144-30
Tantalum carbide ≥99.5% (metals basis)
Tantalum carbide
MDL: MFCD00011255
Purity: 99.5%
CAS No.: 12070-06-3
Molecular Formula: TaC
Formula Weight: 192.96
Physical Form: -325 Mesh Powder
Density: 13.9
Appearance: Grey to black
Odor: Odorless
Melting Point: 3880°
Boiling Point: 5500°
UN No.: 3178
MDL No.: MFCD00011255
Formula:
CTa Boiling Pt: 5500 °C Melting Pt: 3880 °C Density: 13,9 Storage Temperature: Ambient |
MDL Number:
MFCD00011255 CAS Number: 12070-06-3 EINECS: 235-118-3 UN: 3178 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...