Copper(I) cyanide
Supplier: Thermo Scientific Chemicals
|
Danger
|
12135-30
12135-A1
12135-F3
CAAA12135-30EA
91.48
CAD
CAAA12135-30
CAAA12135-A1
CAAA12135-F3
Copper(I) cyanide
Copper (I) cyanide
Electroplating Cu or Fe, polymerization catalyst
CAS No.: 544-92-3
Molecular Formula: CCuN
Formula Weight: 89.56
Physical Form: Powder
Density: 2.92
Appearance: White to cream
Odor: Characteristic
Melting Point: 473°
Solubility: Practically insoluble in water, alcohol. Soluble in NH4OH
Beilstein Reference: 3587244
Merck Reference: 14,2661
UN No.: 1587
MDL No.: MFCD00010975
Electroplating Cu or Fe, polymerization catalyst
Formula:
CCuN Melting Pt: 473 °C Density: 2.92 Storage Temperature: Ambient |
MDL Number:
MFCD00010975 CAS Number: 544-92-3 EINECS: 208-883-6 UN: 1587 ADR: 6.1,II Merck Index: 14,2661 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...