Boron ≥99.5% (metals basis), crystalline pieces ≤ 20 mm (0.8 in)

Supplier: Thermo Scientific Chemicals
Warning

12134-09 12134-18 12134-30
CAAA12134-09EA 203.82 CAD
CAAA12134-09 CAAA12134-18 CAAA12134-30
Boron ≥99.5% (metals basis), crystalline pieces ≤ 20 mm (0.8 in)
Boron

MDL: MFCD00134034


Purity: 99.5% CAS No.: 7440-42-8 Odor: Odorless MDL No.: MFCD00134034

Formula: B
MW: 10.81 g/mol
Density: 1.73 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134034
CAS Number: 7440-42-8
EINECS: 231-151-2
Merck Index: 13,01333

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR