Silver ≥99.998% (metals basis), foil, Premion®, hard, Thickness: 0.1 mm (0.004 in)

Supplier: Thermo Scientific Chemicals

12126-FI 12126-GH 12126-HP
CAAA12126-FIEA 99.7 CAD
CAAA12126-FI CAAA12126-GH CAAA12126-HP
Silver ≥99.998% (metals basis), foil, Premion®, hard, Thickness: 0.1 mm (0.004 in)
Silver

MDL: MFCD00003397


Purity: 99.998% CAS No.: 7440-22-4 Physical Form: ≈2.6g/50x50mm, Temper: as rolled Odor: Characteristic MDL No.: MFCD00003397

Formula: Ag
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 962 °C
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR