Cesium fluoride ≥99.99% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
12121-03
12121-09
12121-18
CAAA12121-03EA
62.57
CAD
CAAA12121-03
CAAA12121-09
CAAA12121-18
Cesium fluoride ≥99.99% (metals basis), Puratronic®
Cesium fluoride
MDL: MFCD00010960
Fieser: 1,121 12,108 13,68 15,75 16,69 17,68 18,88 19,70 20,77 21,95
Purity: 99.99%
CAS No.: 13400-13-0
Molecular Formula: CsF
Formula Weight: 151.90
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
pH: 6.5-7.5
Melting Point: 682°
Boiling Point: 1251°
UN No.: 2923
MDL No.: MFCD00010960
Formula:
CsF MW: 151.9 Boiling Pt: 1251 Melting Pt: 680 Density: 4.627 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010960 CAS Number: 13400-13-0 EINECS: 236-487-3 UN: 2923 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...