Cesium carbonate ≥99.994% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
12117-06
12117-14
12117-22
CAAA12117-06EA
72.36
CAD
CAAA12117-06
CAAA12117-14
CAAA12117-22
Cesium carbonate ≥99.994% (metals basis), Puratronic®
Cesium carbonate
MDL: MFCD00010957
Beilstein Registry No.: 4546405
Purity: 99.994%
CAS No.: 534-17-8
Molecular Formula: Cs2CO3
Formula Weight: 325.82
Physical Form: Powder/Granules
Density: 4.072
pH: 10-12
Melting Point: 610° dec.
Beilstein Reference: 4546405
Merck Reference: 14,2010
MDL No.: MFCD00010957
Formula:
Cs₂CO₃ Melting Pt: 610 °C (dec.) Density: 4.072 Storage Temperature: Ambient |
MDL Number:
MFCD00010957 CAS Number: 534-17-8 EINECS: 208-591-9 Merck Index: 14,2010 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...