Silver (I) sulfide ≥99.995% (metals basis), Premion®
Supplier: Thermo Scientific Chemicals
|
Warning
|
12113-06
12113-14
CAAA12113-06EA
259.12
CAD
CAAA12113-06
CAAA12113-14
Silver (I) sulfide ≥99.995% (metals basis), Premion®
Silver (I) sulphide
MDL: MFCD00003406
Purity: 99.995%
CAS No.: 21548-73-2
Molecular Formula: Ag2S
Formula Weight: 247.80
Physical Form: Powder
Density: 7.317
Appearance: Dark grey to black
Odor: Odorless
Melting Point: 825°
Merck Reference: 14,8530
UN No.: 3077
MDL No.: MFCD00003406
Formula:
Ag₂S Melting Pt: 825 °C Density: 7.317 Storage Temperature: Ambient |
MDL Number:
MFCD00003406 CAS Number: 21548-73-2 EINECS: 244-438-2 UN: 3077 ADR: 9,III Merck Index: 14,8530 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...