Silver sulfate ≥99.999% (metals basis), Premion®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Disilver sulfate, Silver(I) sulfate, Disilver sulphate
12112-06
12112-14
12112-22
CAAA12112-06EA
69.99
CAD
CAAA12112-06
CAAA12112-14
CAAA12112-22
Silver sulfate ≥99.999% (metals basis), Premion®
Silver sulphate
MDL: MFCD00003407
Purity: 99.999%
CAS No.: 10294-26-5
Molecular Formula: Ag2O4S
Formula Weight: 311.80
Physical Form: Powder
Density: 5.45
Appearance: White
Odor: Odorless
Melting Point: 657°
Boiling Point: 1085° dec.
Merck Reference: 14,8529
UN No.: 3077
MDL No.: MFCD00003407
Formula:
Ag₂SO₄ MW: 311.8 g/mol Boiling Pt: 1085 (dec.) Melting Pt: 655 Density: 5.45 Storage Temperature: Ambient |
MDL Number:
MFCD00003407 CAS Number: 10294-26-5 EINECS: 233-653-7 UN: 3077 ADR: 9 III Merck Index: 14,8529 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...