Silver iodide ≥99.999% (metals basis), Premion®
Supplier: Thermo Scientific Chemicals
|
Danger
|
12111-06
12111-14
12111-22
CAAA12111-06EA
131.77
CAD
CAAA12111-06
CAAA12111-14
CAAA12111-22
Silver iodide ≥99.999% (metals basis), Premion®
Silver iodide
MDL: MFCD00003412
Purity: 99.999%
CAS No.: 7783-96-2
Molecular Formula: AgI
Formula Weight: 234.77
Physical Form: Powder
Density: 5.67
Appearance: Yellow
Odor: Odorless
Melting Point: 558°
Merck Reference: 14,8516
UN No.: 3077
MDL No.: MFCD00003412
Formula:
AgI Boiling Pt: 1506 °C (1013 hPa) Melting Pt: 558 °C Density: 5.67 Storage Temperature: Ambient |
MDL Number:
MFCD00003412 CAS Number: 7783-96-2 EINECS: 232-038-0 UN: 3077 ADR: 9,III Merck Index: 14,8516 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...