Silver bromide ≥99.998% (metals basis), Premion®
Supplier: Thermo Scientific Chemicals
12110-06
12110-14
12110-22
CAAA12110-06EA
144.57
CAD
CAAA12110-06
CAAA12110-14
CAAA12110-22
Silver bromide ≥99.998% (metals basis), Premion®
Silver bromide
MDL: MFCD00003398
Purity: 99.998%
CAS No.: 7785-23-1
Molecular Formula: AgBr
Formula Weight: 187.78
Physical Form: Crystalline
Density: 6.47
Appearance: Pale yellow
Odor: Odorless
Melting Point: 432°
Merck Reference: 14,8506
UN No.: 3077
MDL No.: MFCD00003398
Formula:
AgBr Boiling Pt: 1300 °C (1013 hPa) Melting Pt: 432 °C Density: 6.47 Storage Temperature: Ambient |
MDL Number:
MFCD00003398 CAS Number: 7785-23-1 EINECS: 232-076-8 UN: 3077 ADR: 9,III Merck Index: 14,8506 |
Specification Test Results
Form | Crystalline |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...