Zirconium tetrachloride ≥99.5% (metals basis), REacton® reactor grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
12104-09
12104-22
12104-36
12104-A3
CAAA12104-09EA
80.58
CAD
CAAA12104-09
CAAA12104-22
CAAA12104-36
CAAA12104-A3
Zirconium tetrachloride ≥99.5% (metals basis), REacton® reactor grade
Zirconium tetrachloride
Friedel-Crafts catalyst
Purity: 99.5+%
CAS No.: 10026-11-6
Molecular Formula: ZrCl4
Formula Weight: 233.03
Physical Form: Powder
Density: 2.803
Appearance: White to orange
Melting Point: 331° subl.
Solubility: Soluble in alcohol, ether. Decomposes in water forming ZrOCl2 and HCl
Merck Reference: 14,10174
UN No.: 2503
MDL No.: MFCD00011306
Friedel-Crafts catalyst
Formula:
Cl₄Zr Boiling Pt: 3577 °C (1013 hPa) Melting Pt: 331 °C (subl.) Density: 2.803 Storage Temperature: Ambient |
MDL Number:
MFCD00011306 CAS Number: 10026-11-6 EINECS: 233-058-2 UN: 2503 ADR: 8,III Merck Index: 14,10174 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...