Osmium tetraoxide ≥99.8% (metals basis) Os ≥ 74.4%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Osmium (VIII) oxide, Osmic acid anhydride, Osmic acid

12103-02 12103-03 12103-48
CAAA12103-02EA 203.82 CAD
CAAA12103-02 CAAA12103-03 CAAA12103-48
Osmium tetraoxide ≥99.8% (metals basis) Os ≥ 74.4%
Osmium tetraoxide

Oxidation, catalyst in Sharpless dihydroxylations


Purity: 99.8% CAS No.: 20816-12-0 Molecular Formula: OsO4 Formula Weight: 254.10 Physical Form: Crystalline Odor: Like chlorine Boiling Point: 130° Solubility: Soluble in chloroform, alcohol and ethers Merck Reference: 14,6893 UN No.: 2471 MDL No.: MFCD00011150


Oxidation, catalyst in Sharpless dihydroxylations

Formula: O₄Os
MW: 254.23 g/mol
Boiling Pt: 130 °C
Melting Pt: 40.6 °C
Density: 1.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011150
CAS Number: 20816-12-0
EINECS: 244-058-7
UN: 2471
ADR: 6.1,I
Merck Index: 14,6893

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR