Platinum, black

Supplier: Thermo Scientific Chemicals

Synonyms: Platinium

12076-03 12076-06 12076-77
CAAA12076-03EA 357.08 CAD
CAAA12076-03 CAAA12076-06 CAAA12076-77
Platinum, black
Platinum

MDL: MFCD00011179


CAS No.: 7440-06-4 Physical Form: S.A. 10m2/g Odor: Odorless MDL No.: MFCD00011179

Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 4530 °C (1013 hPa)
Melting Pt: 1773 °C
Density: 21.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
UN: 3089
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR