Platinum 99,98 Metals basis, powder -200 mesh

Supplier: Thermo Scientific Chemicals

Synonyms: Platinium

12075-03 12075-06 12075-77
CAAA12075-03EA 575.12 CAD
CAAA12075-03 CAAA12075-06 CAAA12075-77
Platinum 99,98 Metals basis, powder -200 mesh
Platinum

MDL: MFCD00011179


Purity: 99.98% CAS No.: 7440-06-4 Odor: Odorless MDL No.: MFCD00011179

Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
UN: 3089
ADR: 4.1 II
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR