Palladium ≥99.95% (metals basis), powder , -200 mesh

Supplier: Thermo Scientific Chemicals
Warning

12068-03 12068-06 12068-14
CAAA12068-03EA 522.98 CAD
CAAA12068-03 CAAA12068-06 CAAA12068-14
Palladium ≥99.95% (metals basis), powder , -200 mesh
Palladium

MDL: MFCD00011167


Purity: 99.95% CAS No.: 7440-05-3 Appearance: Black Odor: Odorless UN No.: 3089 MDL No.: MFCD00011167

Formula: Pd
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR