Ruthenium ≥99.95% (metals basis), sponge -20 mesh

Supplier: Thermo Scientific Chemicals
Warning

12062-04 12062-09 12062-18
CAAA12062-04EA 376.04 CAD
CAAA12062-04 CAAA12062-09 CAAA12062-18
Ruthenium ≥99.95% (metals basis), sponge -20 mesh
Ruthenium

MDL: MFCD00011207


Purity: 99.95% CAS No.: 7440-18-8 Odor: Odorless UN No.: 3089 MDL No.: MFCD00011207

Formula: Ru
Storage Temperature: Ambient
MDL Number: MFCD00011207
CAS Number: 7440-18-8
EINECS: 231-127-1
UN: 3089
ADR: 4.1,II
Merck Index: 12,08452

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR