Palladium ≥99.99% (metals basis), foil, Premion®, Thickness: 1.0 mm

Supplier: Thermo Scientific Chemicals

12056-FF 12056-FI
CAAA12056-FFEA 3548.68 CAD
CAAA12056-FF CAAA12056-FI
Palladium ≥99.99% (metals basis), foil, Premion®, Thickness: 1.0 mm
Palladium

MDL: MFCD00011167


Purity: 99.99% CAS No.: 7440-05-3 Physical Form: ≈7.5g/25x25mm Odor: Odorless MDL No.: MFCD00011167

Formula: Pd
Boiling Pt: 3167 °C (1013 hPa)
Melting Pt: 1555 °C
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
Merck Index: 12,07121

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR