Zinc ≥99.994% (metals basis), wire, Puratronic®, Ø 0.5 mm (0.02 in)

Supplier: Thermo Scientific Chemicals

12054-G1 12054-G5 12054-G9
CAAA12054-G1EA 75.84 CAD
CAAA12054-G1 CAAA12054-G5 CAAA12054-G9
Zinc ≥99.994% (metals basis), wire, Puratronic®, Ø 0.5 mm (0.02 in)
Zinc

MDL: MFCD00011291


Purity: 99.994% CAS No.: 7440-66-6 Physical Form: ≈1.40g/m Odor: Odorless MDL No.: MFCD00011291

Formula: Zn
Boiling Pt: 908 °C (1013 hPa)
Melting Pt: 420 °C
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR