Lead ≥99.998% (metals basis), foil, Puratronic®, Thickness: 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals
Danger

12050-FI 12050-GH
CAAA12050-FIEA 255.96 CAD
CAAA12050-FI CAAA12050-GH
Lead ≥99.998% (metals basis), foil, Puratronic®, Thickness: 0.25 mm (0.01 in)
Lead

MDL: MFCD00134050


Purity: 99.998% CAS No.: 7439-92-1 Physical Form: ≈7.1g/50x50mm Appearance: Grey Odor: Odorless MDL No.: MFCD00134050

Formula: Pb
Boiling Pt: 1740 °C (1013 hPa)
Melting Pt: 327 °C
Density: 11.3437 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS Number: 7439-92-1
EINECS: 231-100-4
Merck Index: 12,05410

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR