Nickel 99,994 Metals basis, foil, Puratronic®, Thickness: 0.5 mm (0.02 in)

Supplier: Thermo Scientific Chemicals
Danger

12044-FF 12044-FI 12044-GH
CAAA12044-FFEA 192.76 CAD
CAAA12044-FF CAAA12044-FI CAAA12044-GH
Nickel 99,994 Metals basis, foil, Puratronic®, Thickness: 0.5 mm (0.02 in)
Nickel

MDL: MFCD00011137


Purity: 99.994% CAS No.: 7440-02-0 Physical Form: ≈2.8g/25x25mm Odor: Odorless MDL No.: MFCD00011137

Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 3007
Melting Pt: 1453
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR