Palladium ≥99.98%, foil, Thickness: 0.25 mm

Supplier: Thermo Scientific Chemicals

11918-FF 11918-FI
CAAA11918-FFEA 1150.24 CAD
CAAA11918-FF CAAA11918-FI
Palladium ≥99.98%, foil, Thickness: 0.25 mm
Palladium

MDL: MFCD00011167


Purity: 99.98+% CAS No.: 7440-05-3 Physical Form: ≈1.9g/25x25mm Odor: Odorless MDL No.: MFCD00011167

Formula: Pd
Boiling Pt: 3167 °C (1013 hPa)
Melting Pt: 1555 °C
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
Merck Index: 12,07121

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR