Cadmium ≥99.998% (metals basis), wire, Puratronic®, Ø 2.0 mm (0.08 in)

Supplier: Thermo Scientific Chemicals
Danger

11908-BU 11908-G1
CAAA11908-BUEA 126.24 CAD
CAAA11908-BU CAAA11908-G1
Cadmium ≥99.998% (metals basis), wire, Puratronic®, Ø 2.0 mm (0.08 in)
Cadmium

MDL: MFCD00010914


Purity: 99.998% CAS No.: 7440-43-9 Physical Form: ≈27.2g/m MDL No.: MFCD00010914

Formula: Cd
Boiling Pt: 765 °C (1013 hPa)
Melting Pt: 321 °C
Density: 8.642 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010914
CAS Number: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1,II
Merck Index: 12,01649

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR