Tin ≥99.998% (metals basis), wire, Puratronic®, Ø 0.25 mm (0.01 in)
Supplier: Thermo Scientific Chemicals
11907-G1
11907-G5
11907-G9
CAAA11907-G1EA
73.47
CAD
CAAA11907-G1
CAAA11907-G5
CAAA11907-G9
Tin ≥99.998% (metals basis), wire, Puratronic®, Ø 0.25 mm (0.01 in)
Tin
MDL: MFCD00133862
Purity: 99.998%
CAS No.: 7440-31-5
Physical Form: ≈0.36g/m
Odor: Odorless
MDL No.: MFCD00133862
Formula:
Sn Boiling Pt: 2270 °C (1013 hPa) Melting Pt: 232 °C Density: 7.31 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133862 CAS Number: 7440-31-5 EINECS: 23-141-8 Merck Index: 12,09587 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...